Learn More
Invitrogen™ CBR1 Recombinant Rabbit Monoclonal Antibody (24GB3785)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557002
 View more versions of this product
Product Code. 30276640
  • 146.00 € / 20µL
Description
CBR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CBR1
Recombinant Monoclonal
Unconjugated
CBR1
15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1
Rabbit
Affinity chromatography
RUO
12408, 29224, 873
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3785
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P16152, P47727, P48758
CBR1
A synthesized peptide derived from human CBR1 (aa 200-277).
20 μL
Primary
Human, Mouse, Rat