Learn More
CBX1 Antibody (RAB-C145) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20129-0.2mg
 View more versions of this product
Product Code. 18621654
  • 635.00 € / 0.20mg
Description
CBX1 Monoclonal antibody specifically detects CBX1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CBX1
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
RAB-C145
Unconjugated
CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox protein homolog 1, Heterochromatin protein 1 homolog beta, heterochromatin protein 1-beta, Heterochromatin protein p25, heterochromatin protein p25 beta, HP1 beta, HP1 beta homolog, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, p25beta
This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX1 protein under non-denaturing conditions.
0.2 mg
Chromatin Research
10951
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ