missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX1 Antibody (RAB-C145) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-20128-0.1mg
This item is not returnable.
View return policy
Description
CBX1 Monoclonal antibody specifically detects CBX1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Specifications
| CBX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| RAB-C145 | |
| Unconjugated | |
| CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox protein homolog 1, Heterochromatin protein 1 homolog beta, heterochromatin protein 1-beta, Heterochromatin protein p25, heterochromatin protein p25 beta, HP1 beta, HP1 beta homolog, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, p25beta | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX1 protein under non-denaturing conditions. | |
| 0.1 mg | |
| Chromatin Research | |
| 10951 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction