missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CCDA Selective Medium

Product Code. 12982568
Change view
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CCDA Selective Medium
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin unitSize
12982568 10/Pk. 90mm Monoplate CCDA Selective Medium United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12982568 Supplier Thermo Scientific™ Supplier No. PO0119A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Isolate Campylobacter spp. with a blood free selective medium which conforms to ISO 10272-1 and 10272-2.

Isolate Campylobacter spp. using blood-free Thermo Scientific™ CCDA Selective Medium, which conforms to ISO 10272-1 and 10272-2.

Modified CCDA Medium is based on the original formulation described by Bolton which was developed to replace blood with charcoal, ferrous sulphate and sodium pyruvate. Improved selectivity is achieved using cefoperazone as the selective agent. Amphotericin B has been added to the formula to suppress the growth of yeast and fungal contaminants that may occur at 37°C.

Campylobacter infections are among the most common bacterial infections, producing both gastrointestinal and systemic illnesses. Campylobacter jejuni is the most commonly reported bacterial cause of foodborne infection in the United States. Campylobacter are small, non-sporeforming, helical, curved gram-negative rods that require a microaerophilic environment and cannot tolerate drying.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance According ISO 10271-a and ISO 10272-2
Color Black, opaque
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type CCDA Selective Medium
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.