missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC109A Antibody (CL3576) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59029IR
This item is not returnable.
View return policy
Description
CCDC109A Monoclonal antibody specifically detects CCDC109A in Human samples. It is validated for Western Blot,KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| CCDC109A | |
| Monoclonal | |
| DyLight 755 | |
| C10orf42, coiled-coil domain containing 109A, coiled-coil domain-containing protein 109A, FLJ46135 | |
| This antibody was developed using a recombinant protein derived from Q8NE86, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology | |
| 90550 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL3576 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction