missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CCDC109A [p Ser92] Antibody (SR2322), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27827-100ul
This item is not returnable.
View return policy
Description
CCDC109A Monoclonal antibody specifically detects CCDC109A in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| CCDC109A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2322 | |
| Western Blot 1:500-1:2000 | |
| C10orf42, coiled-coil domain containing 109A, coiled-coil domain-containing protein 109A, FLJ46135 | |
| A synthesized peptide derived from human CCDC109A (Uniprot #: Q8NE86) | |
| 100 μL | |
| Cardiovascular Biology, Endocrinology | |
| 90550 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction