missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC137 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12572-100ul
This item is not returnable.
View return policy
Description
CCDC137 Polyclonal antibody specifically detects CCDC137 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| CCDC137 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.5-1uL/mg of uLysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| coiled-coil domain containing 137, coiled-coil domain-containing protein 137, MGC16597 | |
| Produced in rabbits immunized with E. coli-derived Human CCDC137 fragment | |
| 100 μg | |
| Cell Biology | |
| 339230 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction