missing translation for 'onlineSavingsMsg'
Learn More

CCDC27 Monoclonal Antibody (OTI9C6), TrueMAB™, OriGene

Product Code. p-200016860
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505044 30 μL 30µL
17585034 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17505044 Supplier Origene Technologies Supplier No. TA811217S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCDC27 Monoclonal antibody specifically detects CCDC27 in Human samples. It is validated for Western Blot

Specifications

Antigen CCDC27
Applications Western Blot
Classification Monoclonal
Clone OTI9C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NFIA
Gene Accession No. Q12857
Gene Alias CCDC27; coiled-coil domain containing 27; coiled-coil domain-containing protein 27; RP1-286D6.1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 200-509 of human NFIA produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4774
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.