missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC47 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92269-0.1ml
This item is not returnable.
View return policy
Description
CCDC47 Polyclonal antibody specifically detects CCDC47 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CCDC47 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| coiled-coil domain containing 47, coiled-coil domain-containing protein 47, GK001, MSTP041, PSEC0077 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 224-483 of human CCDC47 (NP_064583.2). FLKRQDLLNVLARMMRPVSDQVQIKVTMNDEDMDTYVFAVGTRKALVRLQKEMQDLSEFCSDKPKSGAKYGLPDSLAILSEMGEVTDGMMDTKMVHFLTHYADKIESVHFSDQFSGPKIMQEEGQPLKLPDTKRTLLFTFNVPGSGNTYPKDMEALLPLMNMVIYSIDKAKKFRLNREGKQKADKNRARVEENFLKLTHVQRQEAAQSRREEKKRAEKERIMNEEDPEKQRRLEEAALRREQKKLEKKQMKMKQIKVKAM | |
| 0.1 mL | |
| Cell Biology | |
| 57003 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction