missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC72 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | CCDC72 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18494401
|
Novus Biologicals
NBP2-14594-25ul |
25ul |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18137301
|
Novus Biologicals
NBP2-14594 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC72 Polyclonal specifically detects CCDC72 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CCDC72 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| HSPC016, TMA7 translational machinery associated 7 homolog (S. cerevisiae) | |
| TMA7 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51372 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GSEGGKKKPLKQPKKQAKEMDEEDKAFKQKQKEEPKKLE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title