missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCG-1423, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
CCG-1423 is a novel inhibitor of RhoA/C-mediated gene transcription that is capable of inhibiting invasion of PC-3 prostate cancer cells in a Matrigel model of metastasis.
Specifications
Specifications
| Chemical Name or Material | CCG-1423 |
| Physical Form | Solid |
| CAS | 285986-88-1 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H13ClF6N2O3 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 26 mg/mL (57.17 mM) |
| SMILES | O=C(NOC(C)C(NC1=CC=C(Cl)C=C1)=O)C2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction