missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCKAR Monoclonal Antibody (377251)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561196
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
Specifications
| CCKAR | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P32238 | |
| CCKAR | |
| NS0 mouse myeloma cell line transfected with human CCK-A R, Met1-Gln428. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 377251 | |
| Unconjugated | |
| CCKAR | |
| AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction