missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/MCP1 Antibody (2D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
Brand: Novus Biologicals NBP2-22115SS
This item is not returnable.
View return policy
Description
CCL2/MCP1 Monoclonal specifically detects CCL2/MCP1 in Human, Mouse, Rat, Feline, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CCL2/MCP1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 3 μg/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF | |
| Mouse | |
| 11 kDa | |
| 0.025 mL | |
| Apoptosis, Asthma, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 6347 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2D8 | |
| Unconjugated | |
| P13500 | |
| CCL2 | |
| Purified recombinant fragment of human MCP1 expressed in E. coli. [Uniprot: P13500] | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction