missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/MCP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
234.00 € - 500.00 €
Specifications
| Antigen | CCL2/MCP1 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18465960
|
Novus Biologicals
NBP1-81837 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18446980
|
Novus Biologicals
NBP1-81837-25ul |
25 μL |
234.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL2/MCP1 Polyclonal antibody specifically detects CCL2/MCP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CCL2/MCP1 | |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Asthma, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 6347 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PDAINAPVTCCYNFTNRKISVQRLASYRRITSSKCPKEAVIFKTIVAKEICADPKQKWVQDSMDHLDKQTQTPKT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title