Learn More
CCP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14067-1-AP-20UL
 View more versions of this product
Product Code. 16812544
  • 145.00 € / 20µL
Description
NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.
Specifications
Specifications
CCP1
Polyclonal
Unconjugated
AGTPBP1
AGTPBP1, ATP/GTP binding protein 1, CCP1, Cytosolic carboxypeptidase 1, KIAA1035, NNA1
Rabbit
Antigen Affinity Chromatography
RUO
23287, 67269
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q641K1, Q9UPW5
Agtpbp1
CCP1 Fusion Protein Ag5122
20 μL
Primary
Human, Mouse