missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Antibody (53504) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB145JF585
This item is not returnable.
View return policy
Description
CCR1 Monoclonal antibody specifically detects CCR1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CCR1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Swiss 3T3 mouse cell line transfected with human CCR1, Accession # P32246 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 53504 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1230 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction