Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J328, Paragon Scientific™
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
Specifications
| Packaging | Glass Amber Bottle |
| Certificate of Analysis | Yes |
| Physical Form | Liquid |
| Quantity | 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.