missing translation for 'onlineSavingsMsg'
Learn More

CCT/DRC/ORS Multi-Element Mix 1: Cr, Fe, As, Se @ 100 μg/mL in 2% HNO3, VHG™

Product Code. 30003052
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003052 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003052 Supplier VHG Labs Supplier No. VHGLCELL100
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

CCT/DRC/ORS Multi-Element Mix 1: Cr, Fe, As, Se @ 100 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic/Chromium/Iron/Selenium
CAS 7440-38-2, 7440-47-3, 7439-89-6, 7782-49-2
Methods ICP-MS
Analyte or Component Names Arsenic/Chromium/Iron/Selenium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Arsenic 100 μg/mL/Chromium 100 μg/mL/Iron 100 μg/mL/Selenium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.