missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody

Product Code. 12632177 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Unit Size:
100µL
12mL
1mL
500µL
7mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12632177 500 μL Concentrated 500µL
12622177 100 μL Concentrated 100µL
12612177 1 mL Concentrated 1mL
12692167 7 mL Prediluted 7mL
12602177 12 mL Ready-to-Use 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12632177 Supplier Epredia™ Supplier No. MS728S1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody.

CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.

Host Species: Mouse

Clone: 56C6

Isotype: IgG1

Species Reactivity: Human and Rat. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant external domain of CD10 protein

Molecular Weight: 100kDa

Positive Control: Small intestine

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 56C6
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant external domain of CD10 protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.