missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD103 (Integrin alpha E) Monoclonal Antibody (B-Ly7), NovaFluor™ Red 685, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

185.00 € - 392.00 €

Specifications

Antigen CD103 (Integrin alpha E)
Clone B-Ly7
Concentration 0.4 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055934
View Documents
Invitrogen™
H075T03R02-A
100 Tests
392.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055875
View Documents
Invitrogen™
H075T02R02-A
25 Tests
185.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD103 (Integrin alpha E) Monoclonal antibody specifically detects CD103 (Integrin alpha E) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD103 (Integrin alpha E)
0.4 μg/Test
Monoclonal
Mouse
Human
P38570
3682
Primary
4°C, store in dark, DO NOT FREEZE!
B-Ly7
Flow Cytometry
NovaFluor Red 685
RUO
PBS with 0.09% sodium azide; pH 7.2
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen
IgG1 κ
Affinity chromatography
Itgae
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.