missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 / Integrin alpha E (T-Cell Lymphoma & Hairy Cell Leukemia Marker) Mouse, Clone: ITGAE/2063, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | CD103 / Integrin alpha E (T-Cell Lymphoma & Hairy Cell Leukemia Marker) |
|---|---|
| Clone | ITGAE/2063 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15900288
|
EnquireBio
3682-MSM3-P0 |
20 μg | |||||||
|
15910288
|
EnquireBio
3682-MSM3-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD103 / Integrin alpha E (T-Cell Lymphoma & Hairy Cell Leukemia Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human ITGAE protein fragment | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 150kDa |
| ITGAE/2063 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 3682 | |
| IgG2c κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface and Cytoplasmic | |
| Tonsil, Colon or Stomach |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title