missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD11a/CD18 Antibody (Scripps patent anti-CD11a) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28609-50ug
This item is not returnable.
View return policy
Description
CD11a/CD18 Monoclonal antibody specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD11a/CD18 | |
| Monoclonal | |
| Unconjugated | |
| Q9UKX5 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Scripps patent anti-CD11a | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
| Integrin a11/ ITAG11 | |
| 50 μg | |
| Cytokine Research, Immunology, Signal Transduction | |
| 3683 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction