missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD11a (LFA-1alpha) Monoclonal Antibody (M17/4), NovaFluor™ Red 685, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
185.00 € - 348.00 €
Specifications
| Antigen | CD11a (LFA-1alpha) |
|---|---|
| Clone | M17/4 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055014
|
Invitrogen™
M054T03R02-A |
100 μg |
348.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056292
|
Invitrogen™
M054T02R02-A |
25 μg |
185.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11a (LFA-1alpha) Monoclonal antibody specifically detects CD11a (LFA-1alpha) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD11a (LFA-1alpha) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P24063 | |
| 16408 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| M17/4 | |
| Flow Cytometry | |
| NovaFluor Red 685 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| (p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 | |
| IgG2a κ | |
| Affinity chromatography | |
| ITGAL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title