missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (118/A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00 € - 380.00 €
Specifications
| Antigen | CD11c |
|---|---|
| Clone | 118/A5 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648606
|
Novus Biologicals
NBP2-50130-0.025ml |
0.025 mL |
214.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675276
|
Novus Biologicals
NBP2-50130 |
0.1 mL |
380.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for ImmunohistochemistrySpecifications
| CD11c | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Dendritic Cell Markers, Signal Transduction | |
| PBS | |
| 3687 | |
| IgG2b | |
| Protein A purified |
| 118/A5 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Synthetic peptide conjugated to KLH corresponding to the C terminus of human CD11C: ANGQIAPENGTQTPSPPSEK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title