missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (CL1831), Novus Biologicals™
Mouse Monoclonal Antibody
352.00 € - 572.00 €
Specifications
| Antigen | CD11c |
|---|---|
| Clone | CL1831 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18428361
|
Novus Biologicals
NBP2-34491-25ul |
25 μL |
352.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799333
|
Novus Biologicals
NBP2-34491 |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CD11c Monoclonal antibody specifically detects CD11c in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| CD11c | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| ITGAX | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL1831 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Dendritic Cell Markers, Signal Transduction | |
| P20702 | |
| 3687 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QDGLVDLAVGARGQVLLLRTRPVLWVGVSMQFIPAEIPRSAFECREQVVSEQTLVQSNICLYIDKRSKNLLGSRDLQSSVTLDLALDPGRLSPRATFQETKNRSLSRVRV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title