Learn More
CD11c Armenian Hamster, Biotin, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB23-B-25ug
 View more versions of this product
Product Code. 15911698
  • 116.55 € / 25µg
Specifications
CD11c
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Itgax
Protein A/G Purified
RUO
16411
IgG
Flow Cytometry
N418
Biotin
NP_067309
Armenian Hamster
25 μg
Primary
Store at 2-4°C.