missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Armenian Hamster, PE, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
94.25 € - 240.00 €
Specifications
Antigen | CD11c |
---|---|
Clone | N418 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991698
|
EnquireBio
QAB23-PE-25UG |
25 μg |
94.25 €
25µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981698
|
EnquireBio
QAB23-PE-100UG |
100 μg |
240.00 €
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD11c | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
16411 | |
IgG | |
Protein A/G Purified |
N418 | |
Flow Cytometry | |
PE | |
RUO | |
NP_067309 | |
Itgax | |
Primary | |
Store at 2-4°C. |