missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, Qfluor 710, Clone: 3.9, enQuireBio™
Mouse Monoclonal Antibody
191.00 € - 348.00 €
Especificaciones
Antigen | CD11c |
---|---|
Clone | 3.9 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15901728
|
EnquireBio
QAB24-QF710-25TESTS |
25 Tests |
191.00 €
25 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991718
|
EnquireBio
QAB24-QF710-100TESTS |
100 Tests |
348.00 €
100 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Especificaciones
CD11c | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3687 | |
IgG1 κ | |
Protein A/G Purified |
3.9 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
XP_011544154 | |
ITGAX | |
Primary | |
Store at 2-4°C. |