missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD127 Monoclonal Antibody (eBioRDR5), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 374.00 €
Specifications
| Antigen | CD127 |
|---|---|
| Clone | eBioRDR5 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055144
|
Invitrogen™
H017T03B06-A |
100 Tests | |||||||
|
30054863
|
Invitrogen™
H017T02B06-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow CytometrySpecifications
| CD127 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P16871 | |
| 3575 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioRDR5 | |
| Flow Cytometry | |
| NovaFluor Blue 610-70S | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R α; IL 7Ra; IL 7Rα; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R α; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Rα; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
| IgG1 κ | |
| Affinity chromatography | |
| Il7r |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title