missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD101 Monoclonal Antibody (Moushi101), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ M045T03Y07-A
This item is not returnable.
View return policy
Description
CD101, also known as the V7 antigen, is a transmembrane glycoprotein and a member of the immunoglobulin superfamily. It is characterized by 7 Ig-like IgV domains and a short cytoplasmic tail with several phosphorylation sites. CD101 forms a disulfide-linked homodimer, although its precise function remains unknown. Expression of CD101 is observed on monocytes, granulocytes, dendritic cells, Langerhans cells, and activated T cells. It is also found on a subset of CD25+Foxp3+ regulatory T cells, which exhibit higher suppressive properties in vitro and in vivo. CD101 plays a role in modulating immune responses. Cross-linking CD101 can antagonize TCR/CD3-mediated lymphocyte activation, while CD101+ CD28+ T cells are highly responsive to CD28 signaling. In combination with anti-CD28 or suboptimal levels of anti-CD3, anti-CD101 can enhance proliferation, suggesting an activating role. The monoclonal antibody BB27 has been shown to inhibit T cell reactivity in allogeneic and antigen-specific mixed DC-T cell cultures. In mice, CD101 is expressed on a subpopulation of regulatory T cells (CD4+, CD25+, Foxp3+) with high suppressor activity. In humans, CD101 expression on peripheral blood mononuclear cells (PBMCs) stains approximately 30% of Foxp3+ cells, although studies have not confirmed higher suppressor activity in the human CD101+ population.
Specifications
| CD101 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A8E0Y8 | |
| CD101 | |
| Affinity chromatography | |
| RUO | |
| 630146 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| Moushi101 | |
| NovaFluor Yellow 730 | |
| CD101 | |
| Cd101; CD101 antigen; CD101 molecule; cell surface glycoprotein V7; EWI immunoglobulin subfamily member EWI-101; EWI101; EWI-101; Glu-Trp-Ile EWI motif-containing protein 101; Gm1016; Gm734; IGSF2; Immunoglobulin superfamily member 2; immunoglobulin superfamily, member 2; leukocyte surface protein; RP11-27K13.2; V7 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction