missing translation for 'onlineSavingsMsg'
Learn More

CD134 Mouse anti-Human, BB700, Clone: ACT35 (also known as Ber-ACT35), BD Horizon™

Mouse Monoclonal Antibody

228.31 € - 466.09 €

Specifications

Antigen CD134
Clone ACT35 (also known as Ber-ACT35)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 700 (BB700)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15835149
View Documents
BD Biosciences
566560
25 Tests
ONLINE EXCLUSIVE PRICE
238.00 €
228.31 € / Each
Save 9.69 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825149
View Documents
BD Biosciences
566559
100 Tests
ONLINE EXCLUSIVE PRICE
485.00 €
466.09 € / Each
Save 18.91 €
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD134 Monoclonal antibody specifically detects CD134 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD134
Flow Cytometry
Brilliant Blue 700 (BB700)
Mouse
Human
Primary
ACT35 (also known as Ber-ACT35)
Monoclonal
Purified
RUO
IgG1 κ
Refrigerate (2 to 8°C)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.