missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD134 Rat anti-Mouse, Oligo, Clone: OX-86, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940336
This item is not returnable.
View return policy
Description
CD134 Monoclonal antibody specifically detects CD134 in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD134 | |
| Monoclonal | |
| Oligo | |
| Tnfrsf4; Ox40; OX40L receptor; Ly-70; ACT35; Txgp1; TXGP1L | |
| Recombinant Mouse OX-40 3/4 CD4 Chimeric Protein | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Single Cell 3' Sequencing | |
| Clone OX-86 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction