missing translation for 'onlineSavingsMsg'
Learn More

CD138 Monoclonal Antibody (1D4), PE, Invitrogen™

Product Code. 10705523
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
Product Code. Quantity unitSize
10705523 500 μL Each
1 options
This item is not returnable. View return policy

Product Code. 10705523

Brand: Invitrogen MHCD13804

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.

CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD138
Applications Flow Cytometry
Classification Monoclonal
Clone 1D4
Conjugate PE
Formulation PBS with sucrose and 0.1% sodium azide
Gene SDC1
Gene Accession No. P18827
Gene Alias AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin
Gene Symbols SDC1
Host Species Mouse
Immunogen Human CD138
Purification Method purified
Quantity 500 μL
Regulatory Status ASR
Primary or Secondary Primary
Gene ID (Entrez) 6382
Target Species Human
Content And Storage 4° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.