missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (LPSR/553), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
584.00 € - 631.00 €
Specifications
| Antigen | CD14 |
|---|---|
| Clone | LPSR/553 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667857
|
Novus Biologicals
NBP2-47960B-100ul |
100 μg |
631.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618648
|
Novus Biologicals
NBP2-47960B |
0.1 mL |
584.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| CD14 | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| PBS | |
| 929 | |
| IgG1 κ | |
| Protein A or G purified |
| LPSR/553 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Recombinant full-length human CD14 protein (Uniprot: P08571) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title