missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD14 Monoclonal Antibody (61D3), NovaFluor™ Blue 555, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

182.00 € - 404.00 €

Specifications

Antigen CD14
Clone 61D3
Concentration 0.6 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055106
View Documents
Invitrogen™
H019T03B03-A
100 Tests
404.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055177
View Documents
Invitrogen™
H019T02B03-A
25 Tests
182.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD14
0.6 μg/Test
Monoclonal
Mouse
Human
P08571
929
Primary
4°C, store in dark, DO NOT FREEZE!
61D3
Flow Cytometry
NovaFluor Blue 555
RUO
PBS with 0.09% sodium azide; pH 7.2
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14
IgG1 κ
Affinity chromatography
Cd14
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.