missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD14 Monoclonal Antibody (MEM-15), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
192.00 € - 421.00 €
Specifications
| Antigen | CD14 |
|---|---|
| Clone | MEM-15 |
| Concentration | 0.05 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055865
|
Invitrogen™
H007T03Y06-A |
100 Tests |
421.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055767
|
Invitrogen™
H007T02Y06-A |
25 Tests |
192.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal antibody specifically detects CD14 in Human, Non-human Primate samples. It is validated for Flow CytometrySpecifications
| CD14 | |
| 0.05 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human, Monkey | |
| P08571 | |
| 929 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| MEM-15 | |
| Flow Cytometry | |
| NovaFluor Yellow 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
| IgG1 | |
| Affinity chromatography | |
| Cd14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title