missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD14 Monoclonal Antibody (MEM-15), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H007T03Y07-A
This item is not returnable.
View return policy
Description
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifications
| CD14 | |
| Monoclonal | |
| 0.8 μg/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P08571 | |
| Cd14 | |
| Affinity chromatography | |
| RUO | |
| 929 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG1 |
| Flow Cytometry | |
| MEM-15 | |
| NovaFluor Yellow 730 | |
| Cd14 | |
| CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction