missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, PE, Clone: M5E2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561707

 View more versions of this product

Product Code. 15821259

  • ONLINE EXCLUSIVE PRICE
    81.35 €
    77.19 € / Each
    Save 4.16 €
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD14
Monoclonal
PE
Mouse
Affinity Purified
RUO
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Flow Cytometry
M5E2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD14 Protein
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.