missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse, PE-Cy7, Clone: 61D3, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB26-PE7-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931748

  • 264.00 € / 25 tests
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD14
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD14
Protein A/G Purified
RUO
929
IgG1 κ
Flow Cytometry
61D3
PE-Cyanine7
NP_000582
Mouse
25 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers