missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse, PerCP-Cy5.5, Clone: 61D3, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB26-PCP55-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991738

  • 288.00 € / 25 tests
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD14
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD14
Protein A/G Purified
RUO
929
IgG1 κ
Flow Cytometry
61D3
PerCP-Cyanine5.5
NP_000582
Mouse
25 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers