missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD153 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580170
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: human tonsil tissue. Flow: U937 cell.
The ligand for CD30 is CD30L (CD153). The binding of CD30 to CD30L mediates pleiotropic effects including cell proliferation, activation, differentiation, and apoptotic cell death. CD30 has a critical role in the pathophysiology of Hodgkin's disease and other CD30+ lymphomas. CD30 acts as a costimulatory molecule in thymic negative selection. In addition to its expression on Hodgkin's and Reed-Sternberg cells, CD30 is also found in some non-Hodgkin's lymphomas (including Burkitt's lymphomas), virus-infected T and B cells, and on normal T and B cells after activation. In T cells, CD30 expression is present on a subset of T cells that produce Th2-type cytokines and on CD4+/CD8+ thymocytes that coexpress CD45RO and the IL4 receptor.
Specifications
| CD153 | |
| Polyclonal | |
| Unconjugated | |
| Tnfsf8 | |
| CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 944 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P32971 | |
| Tnfsf8 | |
| E.coli-derived human CD153 recombinant protein (Position: Q63-D234). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur