Learn More
CD154 Armenian Hamster, Biotin, Clone: MR1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB88-B-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15932598
  • 203.00 € / 100µg
Estimated Shipment: 14-06-2024
to see stock.
Specifications
CD154
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd40lg
Protein A/G Purified
RUO
21947
IgG
Flow Cytometry
MR1
Biotin
NP_035746
Armenian Hamster
100 μg
Primary
Store at 2-4°C.