missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), NovaFluor™ Yellow 590, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

178.00 € - 392.00 €

Specifications

Antigen CD157
Clone eBioSY11B5 (SY11B5)
Concentration 0.4 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055085
View Documents
Invitrogen™
H041T03Y02-A
100 Tests
392.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055977
View Documents
Invitrogen™
H041T02Y02-A
25 Tests
178.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD157 Monoclonal antibody specifically detects CD157 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD157
0.4 μg/Test
Monoclonal
Mouse
Human
Q10588
683
Primary
4°C, store in dark, DO NOT FREEZE!
eBioSY11B5 (SY11B5)
Flow Cytometry
NovaFluor Yellow 590
RUO
PBS with 0.09% sodium azide; pH 7.2
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase
IgG1 κ
Affinity chromatography
Bst1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.