missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161/NK1.1 Antibody (PK136) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
208.00 € - 468.00 €
Specifications
| Antigen | CD161/NK1.1 |
|---|---|
| Clone | PK136 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000. Use reported in scientific literature (PMID 21118981), Immunohistochemistry-Frozen 1:20-1:1000 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18779293
|
Novus Biologicals
NB100-77528SS |
0.1 mg |
208.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788103
|
Novus Biologicals
NB100-77528 |
0.5 mg |
468.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD161/NK1.1 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| 17059 | |
| NK-1+ cells from mouse spleen and bone marrow | |
| Primary | |
| Store at 4C. Do not freeze. |
| PK136 | |
| Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000. Use reported in scientific literature (PMID 21118981), Immunohistochemistry-Frozen 1:20-1:1000 | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| Klrb1c | |
| IgG2a κ | |
| Affinity Purified | |
| Mouse Monoclonal to NK-1.1 (PK136) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title