missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 Mouse, Clone: PSGL1/1601, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6404-MSM2-P1
Additional Details : Weight : 0.01000kg
Specifications
CD162 | |
Cell Surface | |
PSGL1/1601 | |
Unconjugated | |
NP_001193538 | |
Mouse | |
43-120kDa | |
Protein A/G Purified | |
RUO | |
6404 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
SELPLG | |
Recombinant human full-length SELPLG (CD162) protein | |
Jurkat cells. Tonsil or Spleen | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |