Learn More
CD163 Antibody (CL10652) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07981JF669
Product Code. 30295160
  • 748.00 € / 0.10mL
Description
CD163 Monoclonal antibody specifically detects CD163 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD163
Monoclonal
Janelia Fluor 669
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
This antibody was developed using a recombinant protein derived from Q86VB7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
9332
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10652
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified