Learn More
CD163 Antibody (EDHu-1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB110-40686AF647
Product Code. 18760594
  • 679.00 € / 0.10mL
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD163
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
Protein A purified
RUO
Primary
NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone.
Human, Pig, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
EDHu-1
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Q86VB7
CD163
Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax.
0.1 mL
Immune System Diseases, Immunology, Innate Immunity
9332
Protein A purified
Store at 4C in the dark.
IgG1