missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (M130/8821R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00 € - 496.00 €
Specifications
| Antigen | CD163 |
|---|---|
| Clone | M130/8821R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682004
|
Novus Biologicals
NBP3-23757-100ug |
100 μg |
496.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616204
|
Novus Biologicals
NBP3-23757-20ug |
20 μg |
268.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD163 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 9332 | |
| IgG κ | |
| Protein A or G purified |
| M130/8821R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Recombinant fragment (around aa 956-1156) of human CD163 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title