missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 (Monocyte & Macrophage Marker) Mouse, Clone: M130/1210, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | CD163 (Monocyte & Macrophage Marker) |
|---|---|
| Clone | M130/1210 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15981198
|
EnquireBio
9332-MSM1-P0 |
20 μg | |||||||
|
15991198
|
EnquireBio
9332-MSM1-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD163 (Monocyte & Macrophage Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human CD163 fragment (apprx. aa 43-196) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 140kDa |
| M130/1210 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 9332 | |
| IgG2b κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface and Cytoplasmic | |
| hPBL Cells. Skin, Lung, Placenta or Histiocytoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title