missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD164L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38010-25ul
This item is not returnable.
View return policy
Description
CD164L2 Polyclonal specifically detects CD164L2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD164L2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q6UWJ8 | |
| CD164L2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VAFFVLHFLKAKDSTYQTLI | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CD164 Sialomucin-Like 2, CD164 Sialomucin-Like 2 Protein, EAPG6122, UNQ6122 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 388611 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction